missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NeutraKine™ IFN-beta Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech 69013-1-IG-100UG
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
The NeutraKine™ IFN beta antibody from Proteintech is a mouse monoclonal antibody generated with recombinantprotein of human NeutraKine™ IFN beta. This antibody recognizes human antigen. The NeutraKine™ IFN beta antibody has been validated for the following applications: ELISA, Neutralization analysis.Specifica
| IFN beta | |
| Monoclonal | |
| Unconjugated | |
| Mouse | |
| Protein G purification | |
| RUO | |
| 3456 | |
| Lyophilized antibodies are stable for 1 year from the date of receipt if stored between (-20°C) and (-80°C). Upon reconstitution we recommend that the solution can be stored at(4°C) for short term or at(-20°C) to (-80°C) for long term. Repeated freeze thaw cycles should be avoided with reconstituted products. | |
| Liquid |
| ELISA, Neutralization | |
| 2C6E5 | |
| IFB; IFF; IFNB; MGC96956 | |
| RecombinantProtein | |
| 100 μg | |
| Primary | |
| Human | |
| Neu Antibody | |
| IgG1 |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto