Learn More
Emma Safety Footwear New York S1P Shoe
Breathable knitted textile for optimal comfort and great performance. With the New York safety shoe, you get optimal protection while looking good.
Marca: Emma Safety Footwear MM43264738
Dettagli aggiuntivi : Peso : 1.06000kg
Descrizione
- A highly sporty shoe with optimum performance in the field of safety. New York is the shoe that combines good looks with maximum protection.
- Thanks to the knitted textile upper and the Nano Carbon Composite protective nose, the New York is particularly light and comfortable. Ideal for workplaces where safety and long-lasting comfort are top priorities.
- The TPU GripForce™ Easy Twist outsole offers maximum grip, is durable and highly recyclable.
- The integrated lacing in this great shoe prevents pressure points from hooks and eyes and the Hydro-Tec™ Antibacterial lining provides optimum breathability.
- New York exudes pure sportsmanship and is designed for performance and looks.
- Industries:
- Logistics & transport
- Electronics
- Manufacturing
- Safety shoe
- S1P-Safetyshoe with sole plate for dry work
Specifica
ISO 20345:2011 | |
Black, Gray, Orange | |
Hydro-Tec Antibacterial | |
38 | |
Standard Width | |
Safety Shoe |
Laces | |
Logistics and Transport, Electronics, Manufacturing | |
Polyurethane | |
Composite | |
High |