missing translation for 'onlineSavingsMsg'
Learn More

NFIA Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marca:  Proteintech 11750-1-AP-20UL

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16899033

  • 129.00€ / 20µL

Spedizione stimata: 23-12-2022
per visualizzare lo stock disponibile
Aggiungi al carrello

Descrizione

Descrizione

11750-1-AP is a rabbit polyclonal antibody raised against N-terminal of human original NFIA.

NF-1, also designated CTF, consists of a family of CCAAT box binding proteins that stimulate DNA replication and activate transcription. Analysis of human NF-1 messenger RNA has revealed two forms of the NF-1 protein arising from an alternate splicing of a single NF-1 gene. NF-1 binds its consensus DNA element as a homodimer via an amino-terminal DNA binding domain, and activates transcription through a putatively novel, proline-rich, carboxy terminal transactivation domain. The NF-1 protein has been shown to recognize and bind the adenovirus type 2 promoter and activate transcription of herpes simplex virus thymidine kinase genes. The NF-1 consensus element has been found in the upstream promoter region of myriad eukaryotic genes, including that of Ha-Ras, alpha-globin, HSP 70, GRP 78, Histone H1, myelin basic protein and in the Xenopus laevis vitellogenin gene promoter.
Specifica

Specifica

NFIA
Polyclonal
Unconjugated
NFIA
CTF, KIAA1439, NF I/A, NF1 A, NFI A, NFI L, NFIA, Nuclear factor 1 A type, Nuclear factor 1/A, nuclear factor I/A, TGGCA binding protein
Rabbit
Antigen Affinity Chromatography
RUO
18027, 25492, 4774
-20°C
Liquid
Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin)
0.37 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P09414, Q02780, Q12857
Nfia
NFIA Fusion Protein Ag2346
20 μL
Primary
Rat, Mouse, Human
Antibody
IgG
Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali