missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NFKB2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 10409-2-AP-20UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
This antibody is a rabbit polyclonal antibody raised against residues near the C terminus of human NFKB2.
The NFkB transcription factor was originally identified as a protein complex consisting of a DNA binding subunit and an associated protein. The subunit is functionally related to c-Rel p75 and Rel B p68. The p50 subunit was initially believed to be a functionally unique protein derived from the amino-terminus of a precursor designated p105. A cDNA has been isolated that encodes an alternative DNA binding subunit of NFkB. It is synthesized as a protein that is expressed in a variety of cell types and, like p105, undergoes cleavage to generate its NFkB subunit, in this case a protein designated p52 (previously referred to as p49). In contrast to p50 derived from p105, p52 acts in synergy with p65 to stimulate the HIV enhancer in transiently transfected Jurkat cells.Specifica
NFKB2 | |
Polyclonal | |
Unconjugated | |
NFKB2 | |
DNA binding factor KBF2, H2TF1, LYT 10, LYT10, NFKB2, NFKB2,p100, NFKB2,p52,p100, Oncogene Lyt 10, p100, p52 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
18034, 309452, 4791 | |
-20°C | |
Liquid |
Western Blot, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.31 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q00653, Q9WTK5 | |
Nfkb2 | |
NFKB2 Fusion Protein Ag0662 | |
20 μL | |
Primary | |
Human, Rat, Mouse | |
Antibody | |
IgG |