missing translation for 'onlineSavingsMsg'
Learn More

NFKB2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marca:  Proteintech 10409-2-AP-150UL

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16896773

  • 399.00€ / 150µL
Spedizione stimata: 20-05-2024
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

This antibody is a rabbit polyclonal antibody raised against residues near the C terminus of human NFKB2.

The NFkB transcription factor was originally identified as a protein complex consisting of a DNA binding subunit and an associated protein. The subunit is functionally related to c-Rel p75 and Rel B p68. The p50 subunit was initially believed to be a functionally unique protein derived from the amino-terminus of a precursor designated p105. A cDNA has been isolated that encodes an alternative DNA binding subunit of NFkB. It is synthesized as a protein that is expressed in a variety of cell types and, like p105, undergoes cleavage to generate its NFkB subunit, in this case a protein designated p52 (previously referred to as p49). In contrast to p50 derived from p105, p52 acts in synergy with p65 to stimulate the HIV enhancer in transiently transfected Jurkat cells.
Specifica

Specifica

NFKB2
Polyclonal
Unconjugated
NFKB2
DNA binding factor KBF2, H2TF1, LYT 10, LYT10, NFKB2, NFKB2,p100, NFKB2,p52,p100, Oncogene Lyt 10, p100, p52
Rabbit
Antigen Affinity Chromatography
RUO
18034, 309452, 4791
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.31 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q00653, Q9WTK5
Nfkb2
NFKB2 Fusion Protein Ag0662
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Suggerimenti di prodotto

Suggerimenti di prodotto

Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali