missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NINJ2 Mouse anti-Human, Clone: 5C5B2, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech 66202-1-IG-150UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
Homophilic cell adhesion molecule that promotes axonal growth. May play a role in nerve regeneration and in the formation and function of other tissues.Specifica
NINJ2 | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9NZG7 | |
NINJ2 | |
NINJ2 Fusion Protein Ag5343 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG2b |
Immunohistochemistry (Paraffin), Western Blot | |
5C5B2 | |
Unconjugated | |
NINJ2 | |
Nerve injury induced protein 2, NINJ2, ninjurin 2 | |
Mouse | |
Protein A | |
RUO | |
4815 | |
-20°C | |
Liquid |