Learn More
Ansell™ NitraSafe™ 28-329 Series Nitrile Heavy Weight Gloves
Click to view available options
Size:
7
8
9
10
Descrizione
- Provides safe handling of oily heavy metal parts, while the NitraSafe™ Foam combines good grip and comfort.
- Both gloves, possibly in combination with the NitraGold™ cuff, offer outstanding mechanical protection against cuts and abrasion.
- Cut and sewn construction
- Cotton Jersey Kevlar™ construction
- Nitrile coating
- Palm-coated finish
Specifica
Materiale parte posteriore | Nitrile |
Certificazione/Conformità | CE Mark 0493, Standard Category III EN 388 : 4331 |
Materiale polso | Nitrile |
Product Type | Heavyweight Gloves |
Size | 10 |
Stile polso guanto | Nitragold™ |
Materiale rivestimento interno | Cotton Jersey Dupont™ Kevlar™ aramid fiber |
Rivestimento interno | Cotton Jersey Dupont™ Kevlar™ aramid fiber |
Tipo di palmo | Coated-palm |
Quantità | 72 Pairs |