Learn More
NKX3-2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca:  Proteintech 22491-1-AP-20UL
 vedi altre versioni di questo prodotto
Codice prodotto. 16831955
  • 145.00€ / 20µL
Questo articolo non è restituibile. Consulta la politica di reso
Description
Description
BAPX1 is the mammalian homolog of the Drosophila bagpipe homeobox gene and is expressed in the splanchnic mesoderm and embryonic skeleton. It is one of the earliest developmental markers for the sclerotome portion of the somite and the gut mesentery. BAPX1 is required for normal skeletal development; homozygous inactivating mutations in the BAPX1 gene result in spodylo-megaepiphyseal-metaphyseal dysplasia (SMMD). It has also been suggested to play a role in the proper development of the mammalian gut and is required for distal stomach development as part of a BARX1-dependent pathway.
Specifications
Specifications
NKX3-2
Polyclonal
Unconjugated
NKX3-2
BAPX1, Homeobox protein Nkx 3.2, NK3 homeobox 2, NKX3 2, NKX3.2, NKX3-2, NKX3B
Rabbit
Antigen Affinity Chromatography
RUO
12020, 579
-20°C
Liquid
Western Blot
0.36 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P78367, P97503
NKX3-2
NKX3-2 Fusion Protein Ag17581
20 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.