missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NMNAT1 Rabbit anti-Human, Mouse, Porcine, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 11399-1-AP-150UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
The coenzyme NAD and its derivatives are involved in hundreds of metabolic redox reactions and are utilized in protein ADP-ribosylation, histone deacetylation, and in some Ca(2+) signaling pathways. NMNAT (EC 2.7.7.1) is a central enzyme in NAD biosynthesis, catalyzing the condensation of nicotinamide mononucleotide (NMN) or nicotinic acid mononucleotide (NaMN) with the AMP moiety of ATP to form NAD or NaAD.The coenzyme NAD and its derivatives are involved in hundreds of metabolic redox reactions and are utilized in protein ADP-ribosylation, histone deacetylation, and in some Ca(2+) signaling pathways. NMNAT (EC 2.7.7.1) is a central enzyme in NAD biosynthesis, catalyzing the condensation of nicotinamide mononucleotide (NMN) or nicotinic acid mononucleotide (NaMN) with the AMP moiety of ATP to form NAD or NaAD.Specifica
NMNAT1 | |
Polyclonal | |
Unconjugated | |
NMNAT1 | |
NaMN adenylyltransferase 1, NMNAT1, PNAT 1, PNAT1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
102160928, 64802, 66454 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9EPA7, Q9HAN9 | |
NMNAT1 | |
NMNAT1 Fusion Protein Ag1944 | |
150 μL | |
Primary | |
Human, Mouse, Porcine | |
Antibody | |
IgG |