missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Merck Novagen™ Lysonase™ Bioprocessing Reagent
Easy, optimized, ready-to-use blend of rLysozyme™ Solution and Benzonase Nuclease. MilliporeSigma™; Lysonase™ Bioprocessing Reagent contains a highly purified and stabilized recombinant lysozyme with specific activity 250 times greater than that of chicken egg white lysozyme.
1285.00€
Specifica
Da utilizzare con (applicazione) | Protein Extraction, Protein Preparation |
---|---|
Product Type | Bioprocessing Reagent |
Content And Storage | -20°C |
Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
---|---|---|---|---|---|---|---|---|---|
Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
17698685
|
Merck
71230-4 |
1 mL |
1285.00€
Pezzo |
Spedizione stimata: 20-06-2024
Eseguire il login per visualizzare lo stock disponibile |
|||||
Descrizione
- Benzonase Nuclease is a genetically engineered nonspecific endonuclease that degrades all forms of DNA and RNA (single stranded, double stranded, circular, linear)
- Reduces extract viscosity and increases protein yield
- Combined activities of rLysozyme and Benzonase Nuclease significantly increase protein extraction efficiency and facilitate downstream processing of protein extracts
- For efficient protein extraction with BugBuster™ Protein Extraction Reagent, use 10μL Lysonase per 1g cell paste
- For efficient protein extraction with PopCulture™ Reagent, add 2μL Lysonase per 1mL culture
Specifica
Protein Extraction, Protein Preparation | |
Bioprocessing Reagent | |
-20°C |