missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NPL Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 16715-1-AP-20UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Catalyzes the cleavage of N-acetylneuraminic acid (sialic acid) to form pyruvate and N-acetylmannosamine via a Schiff base intermediate. It prevents sialic acids from being recycled and returning to the cell surface. Involved in the N- glycolylneuraminic acid (Neu5Gc) degradation pathway. Although human is not able to catalyze formation of Neu5Gc due to the inactive CMAHP enzyme, Neu5Gc is present in food and must be degraded.Specifica
| NPL | |
| Polyclonal | |
| Unconjugated | |
| NPL | |
| c112, C1orf13, N acetylneuraminate lyase, NALase, NPL, Sialate lyase, Sialate pyruvate lyase, Sialic acid aldolase, Sialic acid lyase | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 304860, 74091, 80896 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Western Blot, Immunofluorescence | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q66H59, Q9BXD5, Q9DCJ9 | |
| Npl | |
| NPL Fusion Protein Ag10137 | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto