missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NUDCD3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 11764-1-AP-20UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
NUDCD3 gene ontology annotations related to this gene include cilium assembly; developmental process; protein folding; protein localization to pericentriolar material.Specifica
NUDCD3 | |
Polyclonal | |
Unconjugated | |
NUDCD3 | |
KIAA1068, NudC domain containing 3, NUDCD3, NudCL | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
209586, 23386 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
0.28 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8IVD9, Q8R1N4 | |
Nudcd3 | |
NUDCD3 Fusion Protein Ag2362 | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |