Learn More
NUDT13 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Thermo Scientific PA559234
Dettagli aggiuntivi : Peso : 0.02000kg
Descrizione
Immunogen sequence: QDAQRIEDSV LIGCSEQQEA WFALDLGLDS SFSISASLHK PEMETELKGS FIELRKALFQ LNARDASLLS TAQALLRWH Highest antigen sequence identity to the following orthologs: Mouse - 78%, Rat - 73%.
NUDT13 (Nudix Hydrolase 13) is a Protein Coding gene. Among its related pathways areSynthesis and interconversion of nucleotide di- and triphosphatesandPurine metabolism (REACTOME). Gene Ontology (GO) annotations related to this gene includehydrolase activity.Specifica
NUDT13 | |
Polyclonal | |
Unconjugated | |
NUDT13 | |
4933433B15Rik; LOW QUALITY PROTEIN: nucleoside diphosphate-linked moiety X motif 13; nucleoside diphosphate linked moiety X motif 13; nucleoside diphosphate-linked moiety X motif 13; Nucleoside diphosphate-linked moiety X motif 13-like protein; nudix (nucleoside diphosphate linked moiety X)-type motif 13; nudix hydrolase 13; nudix motif 13; nudix-type motif 13; NUDT13; Protein KiSS-16; Unknown (protein for MGC:140322) | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
25961 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q86X67 | |
NUDT13 | |
Recombinant protein corresponding to Human NUDT13 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only