missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NUDT22 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA551214
Dettagli aggiuntivi : Peso : 0.01000kg
Descrizione
The antibody detects endogenous levels of total NUDT22 protein.
NUDT22 Polyclonal antibody specifically detects NUDT22 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifica
NUDT22 | |
Polyclonal | |
Unconjugated | |
NUDT22 | |
MGC13045; Nucleoside diphosphate-linked moiety X motif 22; NUD22; nudix (nucleoside diphosphate linked moiety X)-type motif 22; nudix hydrolase 22; nudix motif 22; NUDT22; PP11246; UDPG pyrophosphatase; Uridine diphosphate glucose pyrophosphatase NUDT22 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
84304 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.8 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q9BRQ3 | |
NUDT22 | |
Fusion protein of human NUDT22 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |