Learn More
Thermo Scientific™ Nunc™ 384 Well ShallowWell Standard Height Clear Plates
Reduce costs using Thermo Scientific™ Nunc™ 384-well shallow well standard height clear polystyrene plates—total well volume is reduced to 25μL.
Marca: Thermo Scientific™ 264704
Dettagli aggiuntivi : Peso : 3.53430kg
Descrizione
Accommodate small sample volumes by using Thermo Scientific™ Nunc™ 384-Well ShallowWell Standard Height Clear Plates with a total well volume of 25μL, ideal for high throughput screenings. The standard plate height allows for easy adaptation to most automation systems.
Features:- Non-treated hydrophobic polystyrene plates
- ANSI™ (SBS) standard footprint
- Suitable for colorimetric, fluorescent and luminescent assays
- Customized bar coding available
Specifica
All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004) | |
Without Lid | |
16 x 24 | |
Nunc | |
Microplate | |
25 μL | |
2 to 20 μL | |
Non-treated | |
100 |
Standard | |
384 Shallow Well Standard Height, clear, non-treated, no lid, packaged 25/100 | |
100 Cases, 25 Pk. | |
Clear | |
Polystyrene | |
384 | |
Non-sterile | |
Conical Bottom (V-well) | |
25 |