Learn More
NUP133 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Thermo Scientific PA563774
Dettagli aggiuntivi : Peso : 0.02000kg
Descrizione
Immunogen sequence: VTQISVDLMD DYPASDPRWA ESVPEEAPGF SNTSLIILHQ LEDKMKAHSF LMDFIHQVGL FGRLGSFPVR GTPMATRLLL CEHAEKLSAA IVLKNHHSRL SDLVNTAILI ALNKREYEIP SNLTPA Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 83%.
Plays a role in T-cell signaling pathway. Isoform 2 may play a role in T-helper 2 (Th2) signaling pathway and seems to represent the first proximal signaling protein that links T-cell receptor-mediated signal to the activation of c-Maf Th2 specific factor.Specifica
NUP133 | |
Polyclonal | |
Unconjugated | |
NUP133 | |
133 kDa nucleoporin; hNUP133; mermaid; Nuclear pore complex protein Nup133; nucleoporin 133; nucleoporin 133kD; nucleoporin 133kDa; nucleoporin Nup133; NUP133 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55746 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry, Western Blot | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8WUM0 | |
NUP133 | |
Recombinant protein corresponding to Human NUP133 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only