missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NUP85 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 19370-1-AP-20UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
This gene encodes a protein component of the Nup107-160 subunit of the nuclear pore complex. Nuclear pore complexes are embedded in the nuclear envelope and promote bidirectional transport of macromolecules between the cytoplasm and nucleus. The encoded protein can also bind to the C-terminus of chemokine (C-C motif) receptor 2 (CCR2) and promote chemotaxis of monocytes, thereby participating in the inflammatory response. Alternative splicing results in multiple transcript variants.Specifica
NUP85 | |
Polyclonal | |
Unconjugated | |
NUP85 | |
85 kDa nucleoporin, FROUNT, nucleoporin 85kDa, Nucleoporin Nup75, Nucleoporin Nup85, Nup75, NUP85, PCNT1, Pericentrin 1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
445007, 79902 | |
-20°C | |
Liquid |
Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.16 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8R480, Q9BW27 | |
Nup85 | |
NUP85 Fusion Protein Ag7086 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |