Learn More
Cytiva (formerly Pall Lab) Nylaflo™ Nylon Membrane Disc Filters
A highly versatile laboratory membrane filter.
Marca: Cytiva (formerly Pall Lab) 66610
Dettagli aggiuntivi : Peso : 2.16600kg
Descrizione
- Chemical compatibility with esters, bases, and alcohols naturally hydrophilic
- HPLC certified
- Provides assurance that the filter will not add artifacts to your analysis
- For research use only
Specifica
Nylon (Hydrophilic) | |
Non-sterile | |
127 μm |
0.45 μm | |
25/Pk. | |
142 mm |