Learn More
Masterflex™ Raccordo Luer maschio in Nylon
Sterilizable male luer fitting available configured with a variety of fitting options. Cole-Parmer™ Nylon Male Luer Fitting has good chemical resistance.
Marca: Masterflex™ 45505-33
Dettagli aggiuntivi : Peso : 0.01620kg
Descrizione
- Good chemical resistance
- Max temperature 455°F (235°C)
- Sterilize by autoclave, ethylene oxide, or gamma irradiation
Specifica
Raccordo Luer maschio | |
Nylon | |
Da Luer maschio con anello di sicurezza integrato a portagomma da 1/8 in. con foro ampio | |
Conf. da 25 |