Learn More
Motic™ Objective, Achromatic 100X /1.25/S - Oil
Designed with chromatic aberration correction for 2 colors. Motic™ Objective, Achromatic 100X /1.25/ S - Oil is spherically corrected for one color.
Marca: Motic™ 1101001701123
Dettagli aggiuntivi : Peso : 0.01000kg
Descrizione
- Achromatic.
- Chromatic aberration correction for 2 colors.
- Spherical correction for 1 color.
Specifica
Achromat Objective | |
NA: 1.25/S - Oil | |
SFC-100, DM-111, F-1110 Microscopes | |
100X |