Learn More
Zeiss™ Objective EC Plan-Neofluar
Enhanced Contrast Plan-Neofluar universal objectives with consequentliy minimized stray light. Use of glass types of low autofluorescence makes these objectives ideal for epi-fluorescence microscopy
Marca: Zeiss™ 440340-9904-000
Dettagli aggiuntivi : Peso : 0.80000kg
Descrizione
- Visible increase in contrast important for all contrast methods: brightfield, darkfield, phase contrast, DIC, fluorescence, Pol
- Feature high resolving power and brilliant, rich-contrast, completely flat images for observation and documentation
- High transmission of wavelengths from the near UV range (>40% starting at 340nm)
- Special-purpose members of the family are EC Epiplan-Neofluar objectives for reflected-light work
- EC Plan-Neofluar Antiflex objective for reflection contrast
- EC Plan-Neofluar Pol objectives for the requirements of polarizing microscopy
Specifica
Objective EC Plan-Neofluar | |
2 mm | |
20X | |
1 Ea. |