Learn More
Vitlab™ Octagonal Magnetic Stir Bar
Vitlab™ Octagonal Magnetic Stir Bar with rings and permanent magnet AlNiCo V core.
Marca: Vitlab™ 308997
Dettagli aggiuntivi : Peso : 0.01000kg
Descrizione
- The eight-sided shape gives rise to significant turbulence, and thus achieves effective mixing, even at low spin rates
- The middle ring also promotes stable centering with convex or uneven bottoms
Specifica
10 mm | |
35 mm | |
Octagon |