Learn More
Ohaus™ OIML F2 Weight Set, NVLAP Certificate
Supplied with a NVLAP Accredited Certificate and Adjusted to Meet or Exceed OIML F2 Tolerances
Marca:  Ohaus™ 30390848
Dettagli aggiuntivi : Peso : 0.21110kg
Codice prodotto. 16602222
  • 810.00€ / Pezzo
Spedizione stimata: 07-06-2024
per visualizzare lo stock disponibile
Aggiungi al carrello
Descrizione
Descrizione
  • Weights in masses ranging from 1 gram to 1 kilogram are made of Troemner Alloy 8 Stainless Steel which possesses closely controlled density (8.03 g/cm3), extremely low magnetic properties, good stability, and resistance to corrosion
  • Weights are supplied in high quality cases to ensure weights are held securely, kept clean, and dust free
  • NVLAP Calibration Certificate is accredited to ISO 17025 which is recognizable through ILAC
  • Weight surfaces meet or exceed OIML R 111 and have a highly polished mirror-like finish which allows scratches and foreign matters on the weight to be easily seen allowing for stability and longevity
Specifica
Specifica
NVLAP Accredited Certificate
100 g to 1 mg
21/Pk.
Stainless Steel, Aluminum
Protective Case