missing translation for 'onlineSavingsMsg'
Learn More

OMA1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marca:  Proteintech 17116-1-AP-20UL

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16896954

  • 121.38€ / 20µL

Spedizione stimata: 11-10-2022
per visualizzare lo stock disponibile
Aggiungi al carrello

Descrizione

Descrizione

This antibody is specific to OMA1.

OMA1 (Overlapping with the m-AAA protease 1 homolog) is also named as MPRP1(Metalloprotease-related protein 1) and belongs to the peptidase M48 family. It is a zinc metalloprotease that spans the inner mitochondrial membrane with a number of predicted membrane spanning domains and the 60 kDa form of OMA1 rapidly accumulated concomitantly with the cleavage of all OPA1 variants, and the 40 kDa form of OMA1 may be the inactive form, with the 60 kDa form being the active enzyme.
Specifica

Specifica

OMA1
Polyclonal
Unconjugated
OMA1
2010001O09Rik, DAB1, MPRP 1, MPRP1, OMA1, YKR087C, ZMPOMA1
Rabbit
Antigen Affinity Chromatography
RUO
115209, 298282, 67013
-20°C
Liquid
Western Blot, Immunohistochemistry (Paraffin)
0.19 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
D3ZS74, Q96E52, Q9D8H7
Oma1
OMA1 Fusion Protein Ag10803
20 μL
Primary
Rat, Mouse, Human
Antibody
IgG
Videos
SDS
Documenti
Certificati
Offerte speciali

Offerte speciali