Learn More
Kolmi Hopen™ Op-Air™ Type II Medical Mask with Ties and Cellulose Interior Layer
Tie-on medical masks for use in healthcare setting.
Marca: Kolmi Hopen™ M24101-30
6.13 EUR valido fino al 2024-04-30
Usa il codice promo "21673" per avere il tuo prezzo promozionale.
Dettagli aggiuntivi : Peso : 3.42000kg
Descrizione
- High filtration media (polypropylene)
- Layers (interior to exterior): wet laid cellulose, melt blown polypropylene, spunbonded polypropylene
- Soft, non-woven mask ties and edges
- Anatomical folding limits exposure to face
- Integrated nose bar
- Fiberglass-free filter
- Produced in ISO 8 clean room (particle and microbiological control)
Specifica
Blue | |
Polypropylene | |
(EU) 2017/745, EN 14683, ISO 13485, ISO 9001 | |
Medical |
Ties | |
Pleated | |
Yes | |
50 Pk. |