missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ORAI2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 20592-1-AP-150UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Antigen stimulation of immune cells triggers Ca++ entry through Ca++ release-activated Ca++ (CRAC) channels. ORAI2 is one of two mammalian homologs to ORAI1, a recently identified four-transmembrane spanning protein that is an essential component of CRAC. Like ORAI1, ORAI2 has been shown to function as a highly selective Ca++ plasma membrane channel that is gated through interactions with STIM1, the store-activated endoplasmic reticulum Ca++ sensor, although at a lesser efficacy than ORAI1.Specifica
| ORAI2 | |
| Polyclonal | |
| Unconjugated | |
| ORAI2 | |
| C7orf19, CBCIP2, ORAI2, Protein orai 2, TMEM142B, Transmembrane protein 142B | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 269717, 80228 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.17 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8BH10, Q96SN7 | |
| ORAI2 | |
| ORAI2 Fusion Protein Ag14540 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto