Learn More
Thermo Scientific™ Owl™ D2 Wide-Gel Electrophoresis System Combs
Eliminate comb assembly with Thermo Scientific™ Owl™ D2 Wide-Gel Electrophoresis System Combs.
Marca: Thermo Scientific™ D1-10D
Dettagli aggiuntivi : Peso : 0.22000kg
Descrizione
Eliminate comb assembly with Thermo Scientific™ Owl™ D2 Wide-Gel Electrophoresis System Combs. These heavy-duty, one-piece combs are for use with Owl™ D2 Wide-Gel Electrophoresis Systems.
Heavy-duty, one-piece combs eliminate comb assembly.Specifica
Standard; 10 teeth; 1.5mm thick | |
Owl | |
10 |
Owl™ D2 Wide Gel Electrophoresis System | |
Comb | |
1.5 mm |