Learn More
Thermo Scientific™ Owl™ D2 Wide-Gel Electrophoresis System Combs
Eliminate comb assembly with Thermo Scientific™ Owl™ D2 Wide-Gel Electrophoresis System Combs.
Marca:  Thermo Scientific™ D1-40C
Dettagli aggiuntivi : Peso : 0.22000kg
Codice prodotto. 12454597
  • 58.50€ / Pezzo
Spedizione stimata: 30-08-2024
per visualizzare lo stock disponibile
Aggiungi al carrello
Descrizione
Descrizione
Eliminate comb assembly with Thermo Scientific™ Owl™ D2 Wide-Gel Electrophoresis System Combs. These heavy-duty, one-piece combs are for use with Owl™ D2 Wide-Gel Electrophoresis Systems.
Heavy-duty, one-piece combs eliminate comb assembly.
Specifica
Specifica
Standard; 40 teeth; 1.0mm thick
Owl
40
Owl™ D2 Wide Gel Electrophoresis System
Comb
1 mm