missing translation for 'onlineSavingsMsg'
Learn More

P2RX5 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marca:  Proteintech 19012-1-AP-20UL

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16898824

  • 121.38€ / 20µL

Spedizione stimata: 05-10-2022
per visualizzare lo stock disponibile
Aggiungi al carrello

Descrizione

Descrizione

Mmgt2 mediates Mg(2+) transport and may provide regulated pathways for Mg(2+) transport in the Golgi and post-Golgi organelles of epithelium-derived cells. Magnesium is an essential metal, but few selective transporters have been identified. The two separate gene products, Mmgt1 and Mmgt2, comprise the family that we have termed membrane Mg(2+) transporters (MMgTs), and the proteins reside in the membrane and mediate Mg(2+) transport. The transport of Mg(2+) is saturable and studies have identified that Mmgt2 fusion proteins reside in the Golgi complex and post-Golgi vesicles, including the early endosomes in COS-7 cells transfected with the respective tagged constructs. Among related pathways for Mmgt2 are transport and binding events, in particular, transport of glucose and other sugars, bile salts and organic acids, metal ions and amine compounds.
Specifica

Specifica

P2RX5
Polyclonal
Unconjugated
P2RX5
ATP receptor, P2RX5, P2X purinoceptor 5, P2X5, P2X5R, Purinergic receptor
Rabbit
Antigen Affinity Chromatography
RUO
5026
-20°C
Liquid
Western Blot, Immunocytochemistry, Immunofluorescence, Flow Cytometry
0.19 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q93086
P2RX5
P2RX5 Fusion Protein Ag5297
20 μL
Primary
Human
Antibody
IgG
Videos
SDS
Documenti
Certificati
Offerte speciali

Offerte speciali