missing translation for 'onlineSavingsMsg'
Learn More

PAAF1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marca:  Proteintech 17650-1-AP-20UL

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16897584

  • 121.38€ / 20µL

Spedizione stimata: 11-10-2022
per visualizzare lo stock disponibile
Aggiungi al carrello

Descrizione

Descrizione

PAAF1 inhibits proteasome 26S assembly and proteolytic activity by impairing the association of the 19S regulatory complex with the 20S core. In case of HIV-1 infection, recruited by viral Tat to the HIV-1 promoter, where it promotes the recruitment of 19S regulatory complex through dissociation of the proteasome 26S. This presumably promotes provirus transcription efficiency.
Specifica

Specifica

PAAF1
Polyclonal
Unconjugated
PAAF1
PAAF, PAAF1, Protein G 16, Rpn14, WDR71
Rabbit
Antigen Affinity Chromatography
RUO
80227
-20°C
Liquid
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot, Immunofluorescence
0.27 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q9BRP4
PAAF1
PAAF1 Fusion Protein Ag11892
20 μL
Primary
Human, Mouse
Antibody
IgG
Videos
SDS
Documenti
Certificati
Offerte speciali

Offerte speciali