Learn More
PABPC1 PABP Mouse anti-Human, Unconjugated, Clone: 3D8B5, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech 66809-1-IG-150UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
The PABPC1, PABP antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human PABPC1, PABP. This antibody recognizes human, rat antigen. The PABPC1, PABP antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifica
| PABPC1 PABP | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Rat |
| Western Blot, Immunohistochemistry, Immunofluorescence, ELISA | |
| 3D8B5 | |
| Unconjugated | |
| 150 μL | |
| 26986 | |
| IgG1 |