Learn More
Novacyt - Microgen Bioproducts Path-Chek Listeria Swab Test
Simple and instant swab test for the detection of Listeria by direct testing of surfaces.
Marca: Novacyt - Microgen Bioproducts PC080
Dettagli aggiuntivi : Peso : 1.40000kg
Descrizione
- Path-Chek Hygiene Test
- Rapid test system designed for the detection of a range of bacteria from work surfaces and the processing environment.
- Simple and instant swab test for the detection of Listeria by direct testing of surfaces.
Specifica
Listeria species | |
Swab Test | |
Media only (swabs are not included) | |
100 Tests |