Learn More
PBS, Phosphate Buffered Saline, 10X Solution, Fisher BioReagents™
Codice prodotto. 10468543
Click to view available options
Quantità:
500mL
1L
4L
20L
Imballaggio:
Nalgene Poly Bottle
Poly Bottle
Poly Cube
Dimensione della confezione:
1L
20L
4L
500mL
Codice prodotto. 10468543
Marca: Fisher BioReagents BP399500
Richiesta di formato massivo o personalizzato
Tamponi di uso comune
Commonly used buffer
  • Save time and simplify your buffer preparation step by using concentrated Fisher BioReagents 10X PBS solution - simply dilute as needed
  • 10X solution contains 1.37M NaCl, 0.027M KCl and 0.119M phosphates
  • pH of a 1X solution made from this concentrated solution is ∼7.4. To prepare a 1X solution, mix one volume of Fisher BioReagents 10X PBS solution with 9 volumes of ultrapure water, such as BP2819
  • Filtered and tested for the absence of DNase, RNase, and protease
  • PBS is used in many biochemical applications to maintain pH
  • Intended for laboratory research use only
TRUSTED_SUSTAINABILITY
Specifica
Forma fisica Liquid
Quantità 500mL
Imballaggio Poly Bottle