Learn More
Bohlender™ Silicone-lined Screw Caps
Catalog Number 10573873
Click to view available options
Materiale:
TP 220
Constructed of TP220. Kisico™ Silicone-lined Screw Caps are for use with Laboratory Bottles
  • TP220 Construction
  • Silicone-lined
  • GL-18
  • Red
  • Without center hole
Rivestimento interno chiusura Silicone
Materiale chiusura TP 220
Colore Red
Da utilizzare con (applicazione) For closing bottles and vessels with GL threads
Materiale TP 220
Temperature Range (Metric) –42 to 180°C
Dimensioni filettatura (sistema metrico) GL 18