Learn More
PCNA (Proliferating Cell Nuclear Antigen), Rabbit Polyclonal Antibody,Lab Vision™ Epredia™
Provide accurate, reproducible results with the Epredia™ PCNA (Proliferating Cell Nuclear Antigen), Rabbit Polyclonal Antibody.
Marca: Epredia RB-9055-R7
Descrizione
Expression of proliferating cell nuclear antigen (PCNA) or cyclin or polymerase delta auxiliary protein is elevated in the nucleus during late G1 phase immediately before the onset of DNA synthesis, becoming maximal during S-phase and declining during G2 and M phases. PCNA/cyclin may act as an auxiliary protein of DNA polymerase-delta to play a fundamental role in the initiation of cell proliferation.Host Species: Rabbit
Isotype: IgG
Species Reactivity: Human. Others not tested.
Epitope: Not known
Immunogen: Recombinant human PCNA protein
Molecular Weight: 36kDa
Positive Control: Raji cells, Tonsil, Lymph node, and small intestine
Cellular Localization: Nuclear
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
Specifica
| PCNA (Proliferating Cell Nuclear Antigen) | |
| Polyclonal | |
| Rabbit | |
| 7 mL | |
| Cell Cycle | |
| Human | |
| IgG |
| Western Blot | |
| Unconjugated | |
| Recombinant human PCNA protein | |
| IVD | |
| Primary | |
| Prediluted |
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.