missing translation for 'onlineSavingsMsg'
Learn More

PCSK2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marca:  Proteintech 10553-1-AP-20UL

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16897003

  • 121.38€ / 20µL

Spedizione stimata: 11-10-2022
per visualizzare lo stock disponibile
Aggiungi al carrello

Descrizione

Descrizione

The subtilisin-like prohormone convertase (PC) family is a group of cellular enzymes that cleave most prohormones and neuropeptide precursors. Numerous other cellular proteins, some viral proteins, and bacterial toxins that are transported by the constitutive secretory pathway are also targeted for maturation by PCs. PC family members share structural similarities, which include a heterogeneous approximately10 kDa amino-terminal proregion, a highly conserved approximately55 kDa subtilisin-like catalytic domain, and carboxyl-terminal domain that is heterogeneous in length and sequence. These enzymes become catalytically active following proregion cleavage within the appropriate cellular compartment. The subcellular localization of PC family members varies. Immunolocalization studies show that PC1 is found in the perinuclear region as well as the trans-Golgi network, whereas PC2 can be found in the trans-Golgi network as well as diffusely distributed in the peripheral cytoplasm.
Specifica

Specifica

PCSK2
Polyclonal
Unconjugated
PCSK2
KEX2 like endoprotease 2, NEC 2, NEC2, Neuroendocrine convertase 2, PC2, PCSK2, Prohormone convertase 2, Proprotein convertase 2, SPC2
Rabbit
Antigen Affinity Chromatography
RUO
18549, 25121, 5126
-20°C
Liquid
Western Blot
0.13 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P16519, P21661, P28841
PCSK2
PCSK2 Fusion Protein Ag0852
20 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Videos
SDS
Documenti
Certificati
Offerte speciali

Offerte speciali