Learn More
Abnova™ PCSK4 (Human) Recombinant Protein
Human PCSK4 full-length ORF ( AAH36354, 1 a.a. - 242 a.a.) recombinant protein with GST-tag at N-terminal.
Marca: Abnova™ H00054760-P01.25ug
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
- Sequence: MGTRSTLVAIRPLDVSTEGYNNWVFMSTHFWDENPQGVWTLGLENKGYYFNTGTLYRYTLLLYGTAEDMTARPTGPQVTSSACVQRDTEGLCQACDGPAYILGQLCLAYCPPRFFNHTRLVTAGPGHTAAPALRVCSSCHASCYTCRGGSPRDCTSCPPSSTLDQQQGSCMGPTTPDSRPRLRAAACPHHRCPASAMVLSLLAVTLGGPVLCGMSMDLPLYAWLSRARATPTKPQVWLPAGT
Specifica
AAH36354 | |
proprotein convertase subtilisin/kexin type 4 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
PCSK4 | |
GST |
54760 | |
Wheat germ expression system | |
25 μg | |
DKFZp434B217, MGC34749, PC4, SPC5 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |