missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PD-ECGF Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 12383-1-AP-150UL
Questo articolo non è restituibile.
Consulta la politica di reso
Beschreibung
Platelet - derived endothelial growth factor (PD-ECGF), same as thymidine phosphorylase (TP) or gliostatin. In the presence of inorganic orthophosphate, it catalyses the reversible phospholytic cleavage of thymidine and deoxyuridine to their correspondingbases and 2-deoxyribose-1-phosphate. It is both chemotactic and mitogenic for endothelial cells and a non-heparin binding angiogenic factor present in platelets. It is also invoved in transformation of fluoropyrimidines, cytotoxic agents used in the treatment of a variety of maligcies, into active cytotoxic metabolites.Spezifikation
| PD-ECGF | |
| Polyclonal | |
| Unconjugated | |
| TYMP | |
| ECGF1, Gliostatin, hPD ECGF, MNGIE, PD ECGF, PDECGF, TdRPase, thymidine phosphorylase, TP, TYMP | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 1890 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.21 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P19971 | |
| TYMP | |
| PD-ECGF Fusion Protein Ag3052 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur