missing translation for 'onlineSavingsMsg'
Learn More

PDCD10 Mouse anti-Human, Clone: 1E5A8, Proteintech

Mouse Monoclonal Antibody

Marca:  Proteintech 66440-1-IG-20UL

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16898315

  • 121.38€ / 20µL

Spedizione stimata: 03-10-2022
per visualizzare lo stock disponibile
Aggiungi al carrello

Descrizione

Descrizione

This gene encodes an evolutionarily conserved protein associated with cell apoptosis. The protein interacts with the serine/threonine protein kinase MST4 to modulate the extracellular signal-regulated kinase (ERK) pathway. It also interacts with and is phosphoryated by serine/threonine kinase 25, and is thought to function in a signaling pathway essential for vascular developent. Mutations in this gene are one cause of cerebral cavernous malformations, which are vascular malformations that cause seizures and cerebral hemorrhages. Multiple alternatively spliced variants, encoding the same protein, have been identified.
Specifica

Specifica

PDCD10
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9BUL8
PDCD10
PDCD10 Fusion Protein Ag0348
20 μL
Primary
Human
Antibody
IgG1
Immunohistochemistry (Paraffin), Western Blot
1E5A8
Unconjugated
PDCD10
CCM3, PDCD10, programmed cell death 10, TFAR15
Mouse
Protein G
RUO
11235
-20°C
Liquid
Videos
SDS
Documenti
Certificati
Offerte speciali

Offerte speciali