missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PEAK1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA550149
Dettagli aggiuntivi : Peso : 0.01000kg
Descrizione
The antibody detects endogenous levels of total SGK269 / PEAK1 protein.
Tyrosine kinase that may play a role in cell spreading and migration on fibronectin. May directly or indirectly affect phosphorylation levels of cytoskeleton-associated proteins MAPK1/ERK and PXN.Specifica
PEAK1 | |
Polyclonal | |
Unconjugated | |
PEAK1 | |
1110049L02Rik; 9530046P14; AA690169; BE132841; C230081A13Rik; Inactive tyrosine-protein kinase PEAK1; KIAA2002; mKIAA2002; NKF3 kinase family member; Peak1; pseudopodium enriched atypical kinase 1; pseudopodium-enriched atypical kinase 1; Sgk269; sugen kinase 269; Tyrosine-protein kinase SgK269 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
244895, 79834 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q69Z38, Q9H792 | |
PEAK1 | |
Synthesized peptide derived from N-terminal of human SGK269. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |