Learn More
3M™ Pecan Protein ELISA Kit, E96PEC
Intended for the detection of pecan proteins in clean-in-place water (CIP) final rinse water, environmental swab samples, food ingredients and processed food products.
Marca:  3M™ 7100151333
Dettagli aggiuntivi : Peso : 0.02300kg
Codice prodotto. 17213804
  • 899.94€ / 1 set
Spedizione stimata: 29-07-2024
per visualizzare lo stock disponibile
Aggiungi al carrello
Descrizione
Descrizione
  • Designed to detect both processed and unprocessed target allergen proteins.
  • Test can accommodate environmental swab, CIP rinse water and food samples.
  • A universal extraction protocol streamlines testing for multiple analytes, saving you reagents and valuable time.
Specifica
Specifica
Test
96 Wells
65 Minutes