Learn More
Alfa Aesar™ Perfluoroazelaic acid, tech. 90%
Marca: Alfa Aesar™ L16820.14
Dettagli aggiuntivi : CAS : 23453-64-7 Peso : 0.02500kg Transport : UN number : 3261 Chem class : 8 Pack group : II
Quantità | 25g |
---|
Identificatori chimici
23453-64-7 | |
440.089 | |
WRYSWXOLGMKBKW-UHFFFAOYSA-N | |
2776030 | |
C(=O)(C(C(C(C(C(C(C(C(=O)O)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)O |
C9H2F14O4 | |
MFCD00153274 | |
perfluoroazelaic acid, tetradecafluoroazelaic acid, perfluorononanedioic acid, acmc-20ajy5, tetradecafluorononanedioic acid, perfluorononane-1,9-dioic acid, perfluorononane-1,9-dioic acid, tech, 2,2,3,3,4,4,5,5,6,6,7,7,8,8-tetradecakis fluoranyl nonanedioic acid, nonanedioic acid,2,2,3,3,4,4,5,5,6,6,7,7,8,8-tetradecafluoro | |
2,2,3,3,4,4,5,5,6,6,7,7,8,8-tetradecafluorononanedioic acid |
Specifica
Perfluoroazelaic acid | |
25g | |
C9H2F14O4 | |
UN3261 | |
perfluoroazelaic acid, tetradecafluoroazelaic acid, perfluorononanedioic acid, acmc-20ajy5, tetradecafluorononanedioic acid, perfluorononane-1,9-dioic acid, perfluorononane-1,9-dioic acid, tech, 2,2,3,3,4,4,5,5,6,6,7,7,8,8-tetradecakis fluoranyl nonanedioic acid, nonanedioic acid,2,2,3,3,4,4,5,5,6,6,7,7,8,8-tetradecafluoro | |
C(=O)(C(C(C(C(C(C(C(C(=O)O)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)O | |
440.089 | |
440.09 | |
Technical |
215°C (60mmHg) | |
23453-64-7 | |
MFCD00153274 | |
3020403 | |
WRYSWXOLGMKBKW-UHFFFAOYSA-N | |
2,2,3,3,4,4,5,5,6,6,7,7,8,8-tetradecafluorononanedioic acid | |
2776030 | |
90% |