missing translation for 'onlineSavingsMsg'
Learn More

PEX3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marca:  Proteintech 10946-1-AP-150UL

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16897663

  • 325.38€ / 150µL

Spedizione stimata: 06-10-2022
per visualizzare lo stock disponibile
Aggiungi al carrello

Descrizione

Descrizione

Peroxin 3, also known as PEX3 (peroxisomal biogenesis factor 3) or TRG18, is a 373 amino acid multi-pass membrane protein that localizes to peroxisomes and belongs to the Peroxin family. Expressed ubiquitously, Peroxin 3 interacts with Peroxin 19 and is involved in peroxisome biosynthesis and membrane vesicle assembly, as well as in the maintece of peroxisomal integrity. Additionally, Peroxin 3 acts as a docking factor for Peroxin 19 and is required for the import of peroxisomal proteins. Defects in the gene encoding Peroxin 3 are the cause of peroxisome biogenesis disorder complementation group 12 (PBD-CG12) and Zellweger syndrome (ZwS), both of which arise from a failure of peroxisomal protein import.
Specifica

Specifica

PEX3
Polyclonal
Unconjugated
PEX3
Peroxin 3, PEX3, TRG18
Rabbit
Antigen Affinity Chromatography
RUO
56535, 83519, 8504
-20°C
Liquid
Western Blot
0.25 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P56589, Q9JJK4, Q9QXY9
Pex3
PEX3 Fusion Protein Ag1384
150 μL
Primary
Rat, Mouse, Human
Antibody
IgG
Videos
SDS
Documenti
Certificati
Offerte speciali

Offerte speciali