Learn More
Fisherbrand™ pH and Redox Electrode for Mounting Bracket
Maintenance-free
Marca: Fisherbrand™
Dettagli aggiuntivi : Peso : 0.80000kg
Descrizione
- Supplied with 3m cable and BNC plug
- Available in standard and double junction type
- Double junction electrode is suitable for media containing heavy metals, sulphides, substances and Organic compounds
Specifica
BNC Cable | |
3 m | |
Redox Electrode |
Standard | |
0 to 13 |