missing translation for 'onlineSavingsMsg'
Learn More

PHLDA1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marca:  Proteintech 18263-1-AP-20UL

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16898324

  • 121.38€ / 20µL

Spedizione stimata: 28-09-2022
per visualizzare lo stock disponibile
Aggiungi al carrello

Descrizione

Descrizione

Cytotoxic T lymphocyte (CTL)-mediated cytotoxicity constitutes an important component of specific effector mechanisms in immunosurveillance against virus-infected or -transformed cells. Two mechanisms appear to account for this activity, one of which is the perforin-based process. Independently, a FAS-based mechanism involves the transducing molecule FAS (APO-1) and its ligand (FAS-L). The human FAS (APO-1) protein is a 48 kDa cell surface glycoprotein that belongs to a family of receptors that includes CD40, nerve growth factor receptors and tumor necrosis factor receptors. The FAS antigen is expressed on a broad range of lymphoid cell lines, and is expressed at high levels in T cells subsequent to crosslinking of the T cell receptor (TCR). A previously undescribed protein, TDAG51, restores activation- induced apoptosis in cells that have lost the ability to display Fas in response to activation.
Specifica

Specifica

PHLDA1
Polyclonal
Unconjugated
PHLDA1
DT1P1B11, PHLDA1, PHRIP, PQ rich protein, PQR protein, TDAG51
Rabbit
Antigen Affinity Chromatography
RUO
21664, 22822, 29380
-20°C
Liquid
Western Blot, Immunofluorescence, Immunocytochemistry
0.19 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q62392, Q8WV24, Q9QZA1
Phlda1
PHLDA1 Fusion Protein Ag13125
20 μL
Primary
Rat, Mouse, Human
Antibody
IgG
Videos
SDS
Documenti
Certificati
Offerte speciali

Offerte speciali