missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-Akt1 (Ser473) Recombinant Rabbit Monoclonal Antibody (AktS473-B9), FITC, Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Marca: Thermo Scientific MA537324
Dettagli aggiuntivi : Peso : 0.01000kg
Descrizione
Phospho-Akt1 (Ser473) Recombinant Monoclonal antibody specifically detects Phospho-Akt1 (Ser473) in Human, Mouse samples. It is validated for Flow Cytometry.Specifica
Phospho-Akt1 (Ser473) | |
Monoclonal | |
FITC | |
AKT1 | |
Akt; Akt kinase; AKT serine/threonine kinase 1; Akt/PKB; akt1; AKT1 kinase; AKT1m; Akt1m protein; Akt1-PA; Akt1-PB; Akt1-PC; Akt1-PD; Akt1-PE; CAKT; C-AKT; CG4006; CG4006-PA; CG4006-PB; CG4006-PC; CG4006-PD; CG4006-PE; CWS6; dAkt; D-Akt; dAkt kinase; dAKT/dPKB; dAkt/PKB; DAKT1; DAKT1/PKB; Dmel\CG4006; Dmel_CG4006; dPKB; DRAC-PK; DRAC-PK66; DRAC-PK85; l(3)04226; l(3)89Bq; murine thymoma viral (v-akt) oncogene homolog 1; pAkt; P-AKT; PKB; PKB alpha; PKB beta; PKB gamma; PKB/AKT; PKB/dAKT; PKBalpha; PKB-ALPHA; PKBG; PRKBA; protein kinase B; Protein kinase B alpha; protein kinase B-alpha; proto-oncogene c-Akt; RAC; rac protein kinase alpha; RAC protein kinase alpha RAC-PK alpha; RAC serine/threonine protein kinase; RAC serine/threonine-protein kinase; RAC-ALPHA; RAC-alpha serine/threonine-protein kinase; RacPK; RAC-PK-alpha; RAC-PK-beta; RAC-PK-gamma; related to A and C kinases; related to PKA to PKC protein kinases; related to the A and C kinases; SD10374p; serine/threonine protein kinase | |
Rabbit | |
Protein A/G | |
RUO | |
11651, 207 | |
4° C, store in dark | |
Liquid |
Flow Cytometry | |
AktS473-B9 | |
PBS with 0.20% BSA and 0.09% sodium azide | |
P31749, P31750 | |
AKT1 | |
A synthetic phospho-peptide corresponding to residues surrounding Ser473 of human phospho Akt1. | |
10 Tests | |
Primary | |
Human, Mouse | |
Antibody | |
IgG κ |