missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-Akt1 (Ser473) Recombinant Rabbit Monoclonal Antibody (AktS473-C7), PE, Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Marca: Thermo Scientific MA537221
Dettagli aggiuntivi : Peso : 0.01000kg
Description
AKT1 (PKB alpha) is a serine/threonine kinase that regulates cell survival. The activated enzyme inhibits apoptosis and stimulates cell cycle progression by phosphorylating numerous targets in various cell types, including cancer cells. This protein kinase is activated by insulin, PI3K, IGF1 and various other growth and survival factors. Akt promotes cell survival by inhibiting apoptosis through phosphorylation and inactivation of several targets, including forkhead transcription factors, and caspase-9. The AKT pathway is a major target for cancer drug discovery.Specifications
Phospho-Akt1 (Ser473) | |
Monoclonal | |
PE | |
AKT1 | |
Akt; Akt kinase; AKT serine/threonine kinase 1; Akt/PKB; akt1; AKT1 kinase; AKT1m; Akt1m protein; Akt1-PA; Akt1-PB; Akt1-PC; Akt1-PD; Akt1-PE; CAKT; C-AKT; CG4006; CG4006-PA; CG4006-PB; CG4006-PC; CG4006-PD; CG4006-PE; CWS6; dAkt; D-Akt; dAkt kinase; dAKT/dPKB; dAkt/PKB; DAKT1; DAKT1/PKB; Dmel\CG4006; Dmel_CG4006; dPKB; DRAC-PK; DRAC-PK66; DRAC-PK85; l(3)04226; l(3)89Bq; murine thymoma viral (v-akt) oncogene homolog 1; pAkt; P-AKT; PKB; PKB alpha; PKB beta; PKB gamma; PKB/AKT; PKB/dAKT; PKBalpha; PKB-ALPHA; PKBG; PRKBA; protein kinase B; Protein kinase B alpha; protein kinase B-alpha; proto-oncogene c-Akt; RAC; rac protein kinase alpha; RAC protein kinase alpha RAC-PK alpha; RAC serine/threonine protein kinase; RAC serine/threonine-protein kinase; RAC-ALPHA; RAC-alpha serine/threonine-protein kinase; RacPK; RAC-PK-alpha; RAC-PK-beta; RAC-PK-gamma; related to A and C kinases; related to PKA to PKC protein kinases; related to the A and C kinases; SD10374p; serine/threonine protein kinase | |
Rabbit | |
Protein A/G | |
RUO | |
11651, 207 | |
4° C | |
Liquid |
Flow Cytometry | |
AktS473-C7 | |
PBS with 0.20% BSA and 0.09% sodium azide | |
P31749, P31750 | |
AKT1 | |
A synthetic phospho-peptide corresponding to residues surrounding Ser473 of human phospho Akt1. | |
10 Tests | |
Primary | |
Human, Mouse | |
Antibody | |
IgG κ |