missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-Chk1 (Ser317) Recombinant Rabbit Monoclonal Antibody (Chk1S317-F10), APC, Invitrogen™
Rabbit Recombinant Monoclonal Antibody
185.00€ - 466.00€
Specifica
Antigene | Phospho-Chk1 (Ser317) |
---|---|
Clone | Chk1S317-F10 |
Applicazioni | Flow Cytometry |
Classificazione | Monoclonal |
Coniugato | APC |
Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
---|---|---|---|---|---|---|---|---|---|
Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
17196868
|
Thermo Scientific
MA537283 |
10 Tests |
185.00€
pezzi 10 |
Spedizione stimata: 03-06-2024 Eseguire il login per visualizzare lo stock disponibile |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | ||||
17106598
|
Thermo Scientific
MA537004 |
100 Tests |
466.00€
pezzi 100 |
Spedizione stimata: 03-06-2024 Eseguire il login per visualizzare lo stock disponibile |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | ||||
Descrizione
CHEK1 (CHK1) is a serine/threonine protein kinase involved in preventing replication when DNA integrity is compromised. CHK1 is a key mediator in the DNA damage-induced checkpoint network. CHK1 is an evolutionarily conserved protein kinase that functions to ensure genomic integrity upon genotoxic stress. When the G2 or S checkpoint is abrogated by the inhibition of CHK1, p53-deficient cancer cells undergo mitotic catastrophe and eventually apoptosis, whereas normal cells are still arrested in the G1 phase. Thus, CHK1 inhibitors can preferentially potentiate the efficacy of DNA damaging agents in cancer cells, and Chk1 is an attractive therapeutic target for cancer treatment, especially since approximately 50% of all human cancers are p53-deficient.Specifica
Phospho-Chk1 (Ser317) | |
Flow Cytometry | |
APC | |
Rabbit | |
Human, Mouse | |
O14757, O35280 | |
1111, 12649 | |
A synthetic phospho-peptide corresponding to residues surrounding Ser317 of human phospho Chk1. | |
Primary | |
4° C | |
Chek1 |
Chk1S317-F10 | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 0.20% BSA and 0.09% sodium azide | |
C85740; Cell cycle checkpoint kinase; checkpoint kinase 1; checkpoint kinase 1 homolog; checkpoint kinase-1; Checkpoint, S. pombe, homolog of, 1; CHEK1; CHK1; CHK1 checkpoint homolog; Chk1-S; homolog of Chk1-S; HRD1 Protein; rad27; rad27 homolog; serine/threonine protein kinase CHK1; serine/threonine-protein kinase Chk1; Synoviolin; YBR1742; YBR274W | |
Chek1 | |
IgG κ | |
Protein A/G | |
Antibody |