missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-Chk1 (Ser317) Recombinant Rabbit Monoclonal Antibody (Chk1S317-G1), FITC, Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Marca: Thermo Scientific MA537067
Dettagli aggiuntivi : Peso : 0.01000kg
Descrizione
CHEK1 (CHK1) is a serine/threonine protein kinase involved in preventing replication when DNA integrity is compromised. CHK1 is a key mediator in the DNA damage-induced checkpoint network. CHK1 is an evolutionarily conserved protein kinase that functions to ensure genomic integrity upon genotoxic stress. When the G2 or S checkpoint is abrogated by the inhibition of CHK1, p53-deficient cancer cells undergo mitotic catastrophe and eventually apoptosis, whereas normal cells are still arrested in the G1 phase. Thus, CHK1 inhibitors can preferentially potentiate the efficacy of DNA damaging agents in cancer cells, and Chk1 is an attractive therapeutic target for cancer treatment, especially since approximately 50% of all human cancers are p53-deficient.Specifica
Phospho-Chk1 (Ser317) | |
Monoclonal | |
FITC | |
Chek1 | |
C85740; Cell cycle checkpoint kinase; checkpoint kinase 1; checkpoint kinase 1 homolog; checkpoint kinase-1; Checkpoint, S. pombe, homolog of, 1; CHEK1; CHK1; CHK1 checkpoint homolog; Chk1-S; homolog of Chk1-S; HRD1 Protein; rad27; rad27 homolog; serine/threonine protein kinase CHK1; serine/threonine-protein kinase Chk1; Synoviolin; YBR1742; YBR274W | |
Rabbit | |
Protein A/G | |
RUO | |
1111, 12649 | |
4° C, store in dark | |
Liquid |
Flow Cytometry | |
Chk1S317-G1 | |
PBS with 0.20% BSA and 0.09% sodium azide | |
O14757, O35280 | |
Chek1 | |
A synthetic phospho-peptide corresponding to residues surrounding Ser317 of human phospho Chk1. | |
100 Tests | |
Primary | |
Human, Mouse | |
Antibody | |
IgG κ |