missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-CRMP1 (Tyr504) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA564664
Dettagli aggiuntivi : Peso : 0.01000kg
Descrizione
Phospho-CRMP1 (Tyr504) Polyclonal Antibody detects endogenous levels of CRMP1 only when phosphorylated at Tyr504.
Collapsin-response mediator proteins (CRMPs) are highly expressed in the developing brain where they play major roles in axonal outgrowth, neurite differentiation, and apoptosis. Their continued expression in areas of high synaptic remodeling such as the cerebellum, hippocampus, and the olfactory system suggests that these proteins may also be involved in adult brain plasticity. CRMP-1 was initially identified as a dihydro-pyrimidinase expressed exclusively in brain; later studies have shown that it is involved with neurotrophin (NT) 3-induced neurite formation and outgrowth. CRMP-1 localization switches from axonal to somatodendritic when neurons reach functional maturity, suggesting that it is involved in early neuronal differentiation as well as in later processes related to the survival or death of the newly generated neurons.Specifica
Phospho-CRMP1 (Tyr504) | |
Polyclonal | |
Unconjugated | |
CRMP1 | |
Collapsin response mediator protein 1; Crmp1; CRMP-1; dihydropyrimidinase-like 1; dihydropyrimidinase-related protein 1; Dpysl1; DRP1; DRP-1; Ulip3; ULIP-3; Unc-33-like phosphoprotein 3 | |
Rabbit | |
Affinity chromatography | |
RUO | |
12933, 1400, 25415 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P97427, Q14194, Q62950 | |
CRMP1 | |
A synthesized peptide derived from human CRMP-1 (Phospho-Tyr504) | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |